Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017147.2 | NP_058843.1 | 2 | 191 | ||
GenBank mRNA | BC059143.1 | - | 2 | 191 | 6889053 | |
BC086533.1 | - | 2 | 176 | 7305422 | ||
FQ212136.1 | - | 1 | 64 | |||
FQ212794.1 | - | 2 | 190 | |||
FQ212910.1 | - | 2 | 186 | |||
FQ213701.1 | - | 2 | 186 | |||
FQ213897.1 | - | 2 | 215 | |||
FQ214120.1 | - | 2 | 187 | |||
FQ214438.1 | - | 2 | 186 | |||
FQ220571.1 | - | 2 | 188 | |||
FQ226405.1 | - | 2 | 186 | |||
FQ227487.1 | - | 2 | 188 | |||
FQ229073.1 | - | 2 | 215 | |||
FQ229766.1 | - | 2 | 190 | |||
FQ229833.1 | - | 2 | 186 | |||
FQ229875.1 | - | 2 | 190 | |||
FQ230001.1 | - | 2 | 190 | |||
FQ230091.1 | - | 2 | 187 | |||
FQ231379.1 | - | 2 | 186 | |||
FQ232577.1 | - | 1 | 66 | |||
FQ233750.1 | - | 2 | 186 | |||
FQ234162.1 | - | 2 | 190 | |||
FQ234303.1 | - | 2 | 186 | |||
X62908.1 | - | 1 | 115 |