Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srcap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK051146.1 | - | 5 | 952 | ||
AK142089.1 | - | 5 | 996 | |||
AK154145.1 | - | 6 | 637 | |||
AK169954.1 | - | 5 | 980 | |||
BC085144.1 | - | 5 | 935 |