Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100606.1 | NP_001094076.1 | 2 | 277 | ||
GenBank mRNA | BY036102.1 | - | 2 | 277 |