Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089194.1 | - | 1550 | |||
AK129468.1 | - | 1094 | ||||
AK154663.1 | - | 1 | 760 | |||
AK158313.1 | - | 3 | 1455 | |||
AK159547.1 | - | 1627 | ||||
AK170885.1 | - | 1516 | ||||
BC092227.1 | - | 2 | 1405 | 30531788 |