Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014981.1 | NP_001014981.1 | 28 | 5147 | ||
GenBank mRNA | AK122304.1 | - | 19 | 3940 | ||
AK154451.1 | - | 27 | 5031 |