Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142152.1 | - | 10 | 1155 | ||
AK154645.1 | - | 7 | 3395 | |||
AK163238.1 | - | 8 | 2743 | |||
BC031484.1 | - | 10 | 1153 | 2654265 | ||
BC044769.1 | - | 3 | 1098 | |||
BC046438.1 | - | 2 | 129 | |||
BC047372.1 | - | 10 | 1136 | |||
BC057597.1 | - | 8 | 2721 | |||
BC089379.1 | - | 10 | 1110 |