Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267796.1 | NP_001254725.1 | 1 | 470 | ||
GenBank mRNA | AK078053.1 | - | 1 | 470 |