Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033228.3 | NP_001028400.2 | 19 | 2963 | ||
GenBank mRNA | AK035870.1 | - | 19 | 2964 | ||
AK040830.1 | - | 10 | 1387 | |||
AK137169.1 | - | 19 | 2728 | |||
DQ113653.1 | - | 2 | 220 |