Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025837.2 | NP_080113.1 | 5 | 675 | ||
GenBank mRNA | AF244360.1 | - | 5 | 657 | ||
AK003289.1 | - | 3 | 435 | |||
AK003896.1 | - | 5 | 676 | |||
AK132777.1 | - | 5 | 676 | |||
BC009068.1 | - | 5 | 653 | 2650791 | ||
BC051163.1 | - | 5 | 653 | 6466145 |