Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009742.3 | NP_033872.1 | 2 | 717 | ||
GenBank mRNA | BC028762.1 | - | 663 | 1226745 | ||
L16462.1 | - | 2 | 717 |