Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190325.1 | NP_001177254.1 | 3 | 630 | ||
GenBank mRNA | AK137514.1 | - | 2 | 1925 | ||
AK137552.1 | - | 2 | 1925 | |||
M30387.1 | - | 3 | 459 |