Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174099.1 | NP_001167570.1 | 4 | 802 | ||
GenBank mRNA | AK132500.1 | - | 1 | 2165 |