Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190401.2 | NP_001177330.2 | 12 | 1552 | ||
GenBank mRNA | AK148805.1 | - | 10 | 998 | ||
AK149094.1 | - | 12 | 1554 |