Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099635.1 | NP_001093105.1 | 28 | 3801 | ||
GenBank mRNA | AK132623.1 | - | 13 | 2191 | ||
M74753.1 | - | 2 | 181 |