Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020255.3 | NP_064651.2 | 2 | 224 | ||
GenBank mRNA | AF220501.1 | - | 2 | 51 | ||
AK009115.1 | - | 2 | 88 | |||
AK088253.1 | - | 2 | 54 | |||
AK220179.1 | - | 1 | 83 | |||
BC010724.1 | - | 1 | 31 | 2812488 | ||
CA451199.1 | - | 2 | 53 |