Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290283.1 | NP_001277212.1 | 2 | 1089 | ||
NM_183139.2 | NP_898962.2 | 2 | 1085 | |||
GenBank mRNA | AK077214.1 | - | 2 | 1047 | ||
AK139586.1 | - | 2 | 1073 |