Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029469.1 | NP_083745.1 | 1 | 18 | ||
GenBank mRNA | BC125356.1 | - | 1 | 18 | 40129871 | |
BC132637.1 | - | 1 | 18 | 40130914 |