Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Larp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024526.2 | NP_001019697.2 | 16 | 6216 | ||
NM_001080948.2 | NP_001074417.1 | 16 | 6229 | |||
NM_001284521.1 | NP_001271450.1 | 16 | 6203 | |||
NM_001284522.1 | NP_001271451.1 | 16 | 6226 | |||
NM_001284523.1 | NP_001271452.1 | 16 | 6219 | |||
GenBank mRNA | AK087993.1 | - | 1 | 2670 |