Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040459.1 | NP_001035549.1 | 8 | 3908 | ||
GenBank mRNA | AK173129.1 | - | 7 | 3930 | ||
BC151022.1 | - | 8 | 4199 | 9087935 | ||
DQ435686.1 | - | 8 | 3908 | |||
DQ435687.1 | - | 8 | 3908 |