Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kntc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042421.1 | NP_001035886.1 | 44, 45 | 4716 | ||
GenBank mRNA | AK084529.1 | - | 1, 2 | 123 | ||
AK172905.1 | - | 44, 45 | 4690 | |||
BC023686.1 | - | 11, 12 | 1446 |