Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D130043K22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK051381.1 | - | 19 | 3131 | ||
AK122246.1 | - | 20 | 3267 | |||
AK148668.1 | - | 18 | 2874 | |||
BC115723.1 | - | 20 | 3186 | 40086618 | ||
BC115940.1 | - | 20 | 3186 | 40086616 |