Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164225.1 | NP_001157697.1 | 3 | 1334 | ||
GenBank mRNA | BC042620.1 | - | 2 | 1043 | ||
BC132383.1 | - | 2 | 1130 | 40130660 | ||
BC137657.1 | - | 2 | 1130 | 8860676 |