Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098836.1 | NP_001092306.1 | 12 | 1179 | ||
NM_001098837.1 | NP_001092307.1 | 12 | 1158 | |||
GenBank mRNA | AK090368.1 | - | 12 | 1160 | ||
BC059880.1 | - | 6 | 574 | |||
BC130269.1 | - | 11 | 929 | |||
CA315821.1 | - | 2 | 169 |