Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccno | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081062.1 | NP_001074531.1 | 3 | 1385 | ||
GenBank mRNA | AK053890.1 | - | 2 | 1991 | ||
AK086507.1 | - | 3 | 1360 |