Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp13a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128094.1 | NP_001121566.1 | 15, 16 | 1831 | ||
NM_001128096.1 | NP_001121568.1 | 15, 16 | 1831 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY210418.1 | - | 170 | Chr. - - [-] on Build |