Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178923.3 | NP_849254.2 | 20 | 3610 | ||
GenBank mRNA | AK045266.1 | - | 20 | 3598 | ||
AK173120.1 | - | 20 | 3610 | |||
BC023734.1 | - | 17 | 3008 | |||
BC024860.1 | - | 2 | 738 | |||
BC053096.1 | - | 20 | 3427 | 6401143 | ||
BC057592.1 | - | 20 | 3610 | 6417362 |