Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081034.1 | NP_001074503.1 | 13, 14 | 1752 | ||
GenBank mRNA | BC027802.1 | - | 2, 3 | 329 | ||
BC055343.1 | - | 4, 5 | 494 | |||
BC110686.1 | - | 13, 14 | 1491 | |||
BC117884.1 | - | 13, 14 | 1498 | 40096054 | ||
BC117885.1 | - | 13, 14 | 1502 | 40096055 | ||
BC128479.1 | - | 13, 14 | 1502 | 40096053 |