Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033281.3 | NP_001028453.1 | 7 | 1950 | ||
GenBank mRNA | AK133910.1 | - | 7 | 1951 | ||
AK135836.1 | - | 4 | 824 | |||
AK135840.1 | - | 4 | 824 | |||
BC139295.1 | - | 7 | 1980 | 8862317 | ||
DQ340564.1 | - | 7 | 1526 | |||
DQ340565.1 | - | 7 | 1445 |