Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camsap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276359.1 | NP_001263288.1 | 13 | 4312 | ||
NM_001276360.1 | NP_001263289.1 | 14 | 4345 | |||
NM_001276361.1 | NP_001263290.1 | 14 | 4372 | |||
GenBank mRNA | AK035221.1 | - | 6 | 2824 | ||
AK170150.1 | - | 3 | 974 |