Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bahd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045523.1 | NP_001038988.1 | 7 | 3350 | ||
GenBank mRNA | AK129243.1 | - | 7 | 3508 | ||
AK152597.1 | - | 7 | 3290 | |||
BC047433.1 | - | 6 | 2099 | |||
BC100358.1 | - | 7 | 3512 | 30848279 | ||
DV653282.1 | - | 1 | 248 | |||
DV658691.1 | - | 1 | 516 |