Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5c1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085502.1 | NP_001078971.1 | 6 | 775 | ||
GenBank mRNA | BC147308.1 | - | 6 | 824 | 9056382 | |
BC147309.1 | - | 6 | 824 | 9056378 | ||
BC147730.1 | - | 6 | 819 | 9056679 | ||
BC147733.1 | - | 6 | 819 | 9056682 |