Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mboat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126314.2 | NP_001119786.1 | 3 | 984 | ||
GenBank mRNA | BC147844.1 | - | 3 | 970 | 9087561 | |
BC147849.1 | - | 3 | 969 | 9087566 | ||
EU518496.1 | - | 3 | 920 | |||
EU721729.1 | - | 3 | 984 |