Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195437.1 | NP_001182366.1 | 1 | 14 | ||
GenBank mRNA | AI536220.1 | - | 1 | 14 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA123812.1 | - | 14 | Chr. - - [-] on Build |