Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198111.2 | NP_932779.2 | 10 | 3150 | ||
GenBank mRNA | AK147248.1 | - | 10 | 3151 | ||
AK147371.1 | - | 10 | 3151 | |||
BC057114.1 | - | 9 | 3100 | 6405025 | ||
DQ233645.1 | - | 10 | 3206 | |||
DQ233646.1 | - | 9 | 3000 |