Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198111.2 | NP_932779.2 | 14 | 9498 | ||
GenBank mRNA | AK081151.1 | - | 1 | 1734 | ||
AK129115.1 | - | 2 | 4957 |