Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam160b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194345.1 | NP_919326.1 | 14 | 1873 | ||
GenBank mRNA | AK090035.1 | - | 9 | 1124 | ||
BC037228.1 | - | 2 | 150 | |||
BC050861.1 | - | 13 | 1996 | |||
BC058108.1 | - | 2 | 178 | |||
CN529793.1 | - | 5 | 575 |