Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzrn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164593.1 | NP_001158065.1 | 10 | 2602 | ||
NM_001164594.1 | NP_001158066.1 | 8 | 2039 | |||
GenBank mRNA | AK135524.1 | - | 8 | 2022 | ||
AK173342.1 | - | 1 | 952 | |||
BC151034.1 | - | 10 | 2531 | 9087947 |