Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CALM2 SIRNA Assays ›
Gene Symbol:
Calm2
Gene Name:
calmodulin 2
Gene Aliases:
Cam
Chromosome Location:
Chr.6: 11064750 - 11080134 on Build Rnor_6.0
UniGene ID:
Rn.5968
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
50268
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Calm2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
BC058485.1
-
443
6921326
FQ211494.1
-
443
FQ211970.1
-
439
FQ212357.1
-
440
FQ212579.1
-
439
FQ212640.1
-
439
FQ212691.1
-
439
FQ212789.1
-
443
FQ212869.1
-
443
FQ213028.1
-
439
FQ213029.1
-
437
FQ213091.1
-
439
FQ213115.1
-
445
FQ213200.1
-
442
FQ213216.1
-
439
FQ213310.1
-
332
FQ213366.1
-
438
FQ213496.1
-
442
FQ213520.1
-
443
FQ213526.1
-
439
FQ213581.1
-
439
FQ213598.1
-
439
FQ213637.1
-
439
FQ213695.1
-
439
FQ213703.1
-
439
FQ213760.1
-
439
FQ213762.1
-
444
FQ213913.1
-
439
FQ213941.1
-
443
FQ213981.1
-
440
FQ213985.1
-
333
FQ213999.1
-
438
FQ214059.1
-
443
FQ214181.1
-
443
FQ214262.1
-
443
FQ214491.1
-
439
FQ214500.1
-
439
FQ214519.1
-
439
FQ214525.1
-
439
FQ214540.1
-
443
FQ215159.1
-
443
FQ215317.1
-
439
FQ215600.1
-
443
FQ216397.1
-
439
FQ218091.1
-
439
FQ220250.1
-
439
FQ220325.1
-
439
FQ220698.1
-
443
FQ220779.1
-
439
FQ222184.1
-
442
FQ223357.1
-
443
FQ225272.1
-
439
FQ226994.1
-
443
FQ229580.1
-
443
FQ229619.1
-
439
FQ229790.1
-
439
FQ229905.1
-
438
FQ231022.1
-
443
FQ231702.1
-
443
FQ233337.1
-
442
FQ234377.1
-
443
FQ235266.1
-
440
M17069.1
-
441
M19312.1
-
422
Back To Top
More Information
Panther Classification:
Molecular Function -
calmodulin-related
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0