Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033340.3 | NP_001028512.2 | 1 | 821 | ||
GenBank mRNA | AK137598.1 | - | 1 | 822 | ||
BC147419.1 | - | 1 | 764 | 9056500 | ||
BC147420.1 | - | 1 | 764 | 9056487 |