Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sftpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270645.1 | NP_001257574.1 | 5 | 454 | ||
NM_001270647.1 | NP_001257576.1 | 5 | 665 | |||
NM_017329.2 | NP_059025.2 | 5 | 451 | |||
GenBank mRNA | BC085353.1 | - | 5 | 454 | 7121530 | |
CK364490.1 | - | 5 | 677 | |||
M15754.1 | - | 5 | 439 | |||
M33201.1 | - | 5 | 448 | |||
X13176.1 | - | 5 | 439 | |||
X13177.1 | - | 5 | 448 |