Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adnp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175028.1 | NP_778193.1 | 4 | 4001 | ||
GenBank mRNA | AB093268.1 | - | 4 | 4001 | ||
AK088326.1 | - | 1 | 575 | |||
AK160320.1 | - | 1 | 489 | |||
BC024969.1 | - | 1 | 1238 | |||
BC044898.1 | - | 4 | 3906 | |||
BC044904.1 | - | 1 | 1352 | |||
BC064672.1 | - | 2 | 3774 |