Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143956.1 | NP_001137428.1 | 1 | 216 | ||
GenBank mRNA | BC157906.1 | - | 1 | 216 | 9088108 | |
BC158115.1 | - | 1 | 193 | 9088188 | ||
BC172048.1 | - | 1 | 216 | 9054334 | ||
BC172056.1 | - | 1 | 216 |