Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifna15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206870.1 | NP_996753.1 | 1 | 233 | ||
GenBank mRNA | BC116216.2 | - | 1 | 233 | 40091189 |