Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159507.1 | NP_001152979.1 | 24 | 4944 | ||
GenBank mRNA | AK134534.1 | - | 1 | 1491 | ||
BC099701.1 | - | 15 | 3833 |