Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltscr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081418.1 | NP_001074887.1 | 14 | 4640 | ||
GenBank mRNA | AK220278.1 | - | 8 | 5313 |