Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081397.1 | NP_001074866.1 | 36 | 6107 | ||
GenBank mRNA | AB429291.1 | - | 32 | 5739 | ||
AK034257.1 | - | 12 | 3105 | |||
BC082311.1 | - | 35 | 6099 | |||
BC151049.1 | - | 35 | 6187 | 9087962 | ||
BC151051.1 | - | 35 | 6188 | 9087964 |