Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgr15l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033361.3 | NP_001028533.1 | 5 | 4485 | ||
GenBank mRNA | AK136917.1 | - | 5 | 4416 | ||
AK138339.1 | - | 5 | 4485 |
Set Membership: |
G-protein coupled receptor |