Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080549.2 | NP_001074018.1 | 3 | 489 | ||
NM_001290459.1 | NP_001277388.1 | 2 | 342 | |||
GenBank mRNA | AK083436.1 | - | 2 | 330 | ||
BC116300.1 | - | 3 | 432 | 40095063 | ||
BC116301.1 | - | 2 | 285 | 40095064 |