Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012018.2 | NP_036148.2 | 40 | 6753 | ||
GenBank mRNA | AK084186.1 | - | 1 | 23 | ||
AK131139.1 | - | 18 | 3107 | |||
BC079536.1 | - | 28 | 4778 | |||
BC158010.1 | - | 39 | 6626 | 9007318 |