Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081162.1 | NP_001074631.1 | 4 | 352 | ||
GenBank mRNA | AK052665.1 | - | 4 | 303 | ||
AK158115.1 | - | 4 | 306 | |||
BC111884.1 | - | 3 | 147 | 40060299 |